Learn More
Cytiva HiTrap Albumin and IgG Depletion
HiTrap Albumin and IgG Depletion is a prepacked columns for the depletion of albumin and IgG from human serum and plasma. The columns are prepacked with high performance SepharoseTM based media with an affinity for human serum albumin (HSA) and IgG
Brand: Cytiva 28-9466-03
Code : 50
Additional Details : Weight : 0.08500kg
Description
- HiTrap Albumin and IgG Depletion is a prepacked column for the depletion of albumin and IgG from human serum or plasma.
- HiTrap Albumin and IgG Depletion column provides rapid and easy processing in a convenient format for handling larger sample volumes (∼150 μl) using a liquid chromatography system or manually using a syringe.
- Columns are prepacked with high performance Sepharose based media with an affinity for human serum albumin (HSA) and IgG.
- High depletion capacity, removes > 95% HSA and > 90% IgG, with high reproducibility.
- Simple and fast procedure.
- Removal of albumin and IgG allows a higher load of less abundant proteins to be included in the analysis and detection of more proteins.
Specifications
7 mm | |
25 mm | |
34 μm | |
1 mL | |
Polypropylene |
<4 mL/min | |
Recombinant Protein G fragment and recombinant Protein binding HAS | |
3 to 9 | |
No | |
Guard Cartridge |
Safety and Handling
- HiTrap Albumin & IgG Depletion
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.