missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ HistoStar™ Heated Forceps

Product Code. 12628216 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Heated Forceps, 1 mm tip
Heated Forceps, 2 mm tip
Heated Forceps, 4 mm tip
Color:
Blue
Red
Yellow
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Color unitSize
12628216 Heated Forceps, 2 mm tip Yellow Each
12638216 Heated Forceps, 4 mm tip Blue Each
12618216 Heated Forceps, 1 mm tip Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12628216 Supplier Epredia™ Supplier No. A81010034
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Epredia™ Heated Forceps work in conjunction with the Epredia™ HistoStar™ Embedding Workstation. Multiple sizes are available.

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
For Use With (Equipment) Epredia™ HistoStar™ Embedding Workstation
Product Type Heated Forceps, 2 mm tip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.