missing translation for 'onlineSavingsMsg'
Learn More

Histone, MP Biomedicals™

Product Code. 11393909 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393909 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11393909 Supplier MP Biomedicals Supplier No. 0210072650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Commonly used for chromosome-reconstitution studies. Histone, MP Biomedicals is an arginine-rich subgroup that is a suitable substrate for protein kinase C.

TRUSTED_SUSTAINABILITY

Specifications

CAS 37244-51-2
Quantity 50 mg
Physical Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.