missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H4, Trimethyl Lys20 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£257.00 - £400.00
Specifications
| Antigen | Histone H4 (Trimethyl Lys20) |
|---|---|
| Concentration | 0.58 mg/mL |
| Dilution | Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry reported in scientific literature (PMID 23924899), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23924899), Dot Blot 0.5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Dot Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18230701
|
Novus Biologicals
NB21-2090SS |
0.025 mg |
£257.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18259450
|
Novus Biologicals
NB21-2090 |
0.05 mg |
£400.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H4 Polyclonal specifically detects Histone H4 in Human, Mouse, Rat, Mammal samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Dot Blot.Specifications
| Histone H4 (Trimethyl Lys20) | |
| Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry reported in scientific literature (PMID 23924899), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23924899), Dot Blot 0.5 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat, Mammalia | |
| P62805, P62806 | |
| 121504 | |
| Synthetic trimethylated peptide surrounding Lysine 20 of human Histone H4 [Swiss Prot# P62805]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.58 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Dot Blot | |
| Unconjugated | |
| RUO | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4K20Me3, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
| HIST4H4 | |
| IgG | |
| Affinity Purified | |
| Human, mouse and rat. Predicted to react with most mammal species. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title