missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H4, p Ser1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB21-2000
This item is not returnable.
View return policy
Description
Histone H4 Polyclonal specifically detects Histone H4 in Human, Mouse, Rat, Mammal samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Dot Blot.
Specifications
| Histone H4 (p Ser1) | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4S1p, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 121504 | |
| Human, Mouse, Rat, Mammalia | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Dot Blot, ChIP Assay | |
| 0.89 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Dot Blot 0.1 ug/ml, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells | |
| P62805, P62806 | |
| HIST4H4 | |
| Synthetic phosphorylated peptide surrounding Serine 1 of human Histone H4 [Swiss Prot# P62805]. | |
| 0.05 mg | |
| Primary | |
| Human, mouse and rat. Predicted to react with most mammal species. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction