missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£173.00 - £302.00
Specifications
| Antigen | Histone H4 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18061154
|
Novus Biologicals
NB100-56348SS |
0.025 mg |
£173.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18076854
|
Novus Biologicals
NB100-56348 |
0.1 mg |
£302.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H4 Polyclonal specifically detects Histone H4 in Human samples. It is validated for Western Blot.Specifications
| Histone H4 | |
| Western Blot 2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
| HIST4H4 | |
| IgG | |
| Protein G purified | |
| A 14 kDa band is observed. The has been evaluated in human, and most likely reacts with additional mammalian, invertebrate, and plant species as the immunogen sequence used to make it is conserved. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, Histones and Modified Histones | |
| PBS with 0.05% Sodium Azide | |
| 121504 | |
| This antibody was generated by immunizing rabbits with a synthetic peptide containing amino acid residues 15-30 of human Histone H4 GAKRHRKVLRDNIQGI. This sequence is conserved across mammalian, invertebrate and plant species. An NCBI BLAST shows no cross-reactivity of this sequence with other known histones. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title