missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H4, ac Lys16 Antibody (C.15200219), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59242
This item is not returnable.
View return policy
Description
Histone H4 Monoclonal specifically detects Histone H4 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Dot Blot.
Specifications
| Histone H4 (ac Lys16) | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| HIST4H4 | |
| The exact immunogen is propietary information. | |
| 50 μg | |
| Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones | |
| 121504 | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Dot Blot, ChIP Assay | |
| C.15200219 | |
| Western Blot 1:500, Dot Blot 1:10000, Chromatin Immunoprecipitation (ChIP) 1-2 ug | |
| H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4K16ac, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction