missing translation for 'onlineSavingsMsg'
Learn More

Histone H4, ac Lys12, ac Lys8, ac Lys16 Antibody (KM-2), DyLight 594, Novus Biologicals™

Product Code. 18365843 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365843 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365843 Supplier Novus Biologicals Supplier No. NBP312018DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Histone H4 Monoclonal antibody specifically detects Histone H4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H4
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone KM-2
Conjugate DyLight 594
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Host Species Mouse
Immunogen KM-2 antibody was derived from an MRL/lpr lupus mouse.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 121504
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.