missing translation for 'onlineSavingsMsg'
Learn More

Histone H3, Trimethyl Lys9 Antibody (6F12-H4), Novus Biologicals™

Product Code. p-4982334 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18715383 0.025 mL 25µL
18731484 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18715383 Supplier Novus Biologicals Supplier No. NBP130141SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 19 publications

Histone H3 Monoclonal specifically detects Histone H3 in Human, Mouse, Rat, Porcine, C. elegans, Drosophila, Invertebrate, Mammal, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3 (Trimethyl Lys9)
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 6F12-H4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:1000. Use reported in scientific literature (PMID 23872316), Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 28270554), Immunohistochemistry-Paraffin 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Gene Accession No. P84243, P84244
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Mouse
Immunogen This Histone H3 [Trimethyl Lys9] antibody (6F12-H4) was raised against a synthetic peptide made to an N-terminal region of Histone H3 (between amino acids 1-50). [UniProt# P84243]
Molecular Weight of Antigen 15 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Test Specificity All mammals including human, mouse and rat, Drosophila, C. elegans, yeast. Invertebrate/Blattella germanica (German cockroach) reactivity reported in scientific literature (PMID: 23872316).
Target Species Human, Mouse, Rat, Pig, C. elegans, Drosophila, Invertebrate, Mammalia, Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.