missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, Trimethyl Lys4 Antibody (RM340), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-77403
This item is not returnable.
View return policy
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Vertebrate samples. It is validated for ELISA, ChIP assay (ChIP), Multiplex Immunoassay
Specifications
| Histone H3 | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1 μg/mL - 10 μg/mL, Chromatin Immunoprecipitation (ChIP) 1 - 5 μg/mL, Multiplex Immunoassay 0.5 μg/mL - 2 μg/mL | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K4Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| This Histone H3 [Trimethyl Lys4] antibody (RM340) was raised against a trimethyl-peptide corresponding to Histone H3 [Trimethyl Lys4] | |
| 100 μg | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, ChIP Assay, Multiplex | |
| RM340 | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Vertebrates | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction