missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, Trimethyl Lys4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
£250.00 - £406.00
Specifications
| Antigen | Histone H3 (Trimethyl Lys4) |
|---|---|
| Concentration | 0.7 mg/ml |
| Dilution | Western Blot 1 ug/ml, Immunohistochemistry reported in scientific literature (PMID 33921015), Immunocytochemistry/Immunofluorescence 1:500, Dot Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Dot Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18069745
|
Novus Biologicals
NB21-1023SS |
0.025 mg |
£250.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18774943
|
Novus Biologicals
NB21-1023 |
£406.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans, Chicken, Xenopus samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Dot Blot.Specifications
| Histone H3 (Trimethyl Lys4) | |
| Western Blot 1 ug/ml, Immunohistochemistry reported in scientific literature (PMID 33921015), Immunocytochemistry/Immunofluorescence 1:500, Dot Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells | |
| Polyclonal | |
| Rabbit | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| PBS and 30% Glycerol with 0.01% Sodium Azide | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K4Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| H3C14 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant (Nicotiana tabacum) based on 100% sequence homology. |
| 0.7 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Dot Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, C. elegans, Chicken, Xenopus species | |
| Q71DI3, P84228 | |
| 126961 | |
| This Histone H3 [Trimethyl Lys4] antibody was raised against synthetic trimethylated peptide surrounding Lysine 4 of human Histone H3.2 [Swiss Prot Q71DI3]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 15 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title