missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, Trimethyl Lys27 Antibody (BT164), DyLight 650, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
| Antigen | Histone H3 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | BT164 |
| Conjugate | DyLight 650 |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 |
| Host Species | Mouse |
| Immunogen | BT164 is a mice-derrived lupus antibody that was established by selection on apoptotic chromatin. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?