missing translation for 'onlineSavingsMsg'
Learn More

Histone H3, Trimethyl Lys27 Antibody (BT164), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18319871
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319871 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319871 Supplier Bio-Techne Supplier No. NBP312024AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Histone H3 Monoclonal antibody specifically detects Histone H3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone BT164
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Host Species Mouse
Immunogen BT164 is a mice-derrived lupus antibody that was established by selection on apoptotic chromatin.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.