missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, Trimethyl Lys20 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54620
This item is not returnable.
View return policy
Description
Histone H3 Polyclonal antibody specifically detects Histone H3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Dot Blot, Protein Array, ChIP assay (ChIP), ChIP assay Sequencing
Specifications
| Histone H3 | |
| Polyclonal | |
| Western Blot 1:1000, ELISA 1:500, Immunocytochemistry/ Immunofluorescence 1:500, Dot Blot 1:20000, Protein Array 1:10000, Chromatin Immunoprecipitation (ChIP) 0.5-1 μg/IP, Chromatin Immunoprecipitation Sequencing | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K20Me3, HIST2H3C, Histone Cluster 3 H3, histone H3.3, MGC87782, MGC87783 | |
| This Histone H3 [Trimethyl Lys20] antibody was developed against H4K20Me3 | |
| 50 μg | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence, Dot Blot, Protein Array | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction