missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, p Thr6 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB21-1051G
This item is not returnable.
View return policy
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Histone H3 (p Thr6) | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Q71DI3 | |
| H3C14 | |
| This Histone H3 [p Thr6] antibody was raised against synthetic peptide surrounding Threonine 6 of human Histone H3.2 [Swiss Prot# Q71DI3]. | |
| 0.05 mL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Human, Mouse, C. elegans | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction