missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, Monomethyl Lys37 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB21-1271-0.05MG
This item is not returnable.
View return policy
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Dot Blot, ICC/IF (Negative), Chromatin Immunoprecipitation (Negative).
Specifications
| Histone H3 (Monomethyl Lys37) | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| H3 histone, family 3A, H3.3AH3F3H3F3B, H3.3B, histone H3.3, MGC87782, MGC87783 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, C. elegans | |
| IgG |
| ChIP Assay, Dot Blot, Immunocytochemistry | |
| 0.42 mg/mL | |
| Western Blot 1ug/ml, Dot Blot 1:1000, ICC/IF (Negative), Chromatin Immunoprecipitation (Negative) | |
| Q71DI3, P84228 | |
| HIST2H3C | |
| Synthetic methylated peptide surrounding Lysine 37 of human Histone H3 [Swiss Prot Q71DI3]. | |
| 0.05 mg | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones | |
| 126961 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction