missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, Dimethyl Lys9 Antibody (C.15200154), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59294
This item is not returnable.
View return policy
Description
Histone H3 (Dimethyl Lys9) Monoclonal antibody specifically detects Histone H3 (Dimethyl Lys9) in Human, Plant, Zebrafish samples. It is validated for Western Blot, ChIP assay, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications
| Histone H3 (Dimethyl Lys9) | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| H3C14 | |
| The exact sequence of the immunogen to this Histone H3 [Dimethyl Lys9] antibody (C.15200154) is proprietary. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Plant, Zebrafish | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay | |
| C.15200154 | |
| Western Blot 1:1000, ELISA 1:1000, Immunocytochemistry/Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 3 ug | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9Me2, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Mouse | |
| 15 kDa | |
| 50 μg | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction