missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3 [Dimethyl Lys4] Antibody (JE00-98), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32431
This item is not returnable.
View return policy
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human,Monkey,Mouse,Rat samples. It is validated for ChIP assay,Western Blot,Dot Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Histone H3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Chromatin Immunoprecipitation Use 0.5-2 μg for 25 μg of chromatin, Immunocytochemistry/Immunofluorescence 1:500-1:2000, Dot Blot 1:1000 | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Synthetic peptide corresponding to the amino terminus of histone H3 in which Lys4 is di-methylated. (Uniprot: P68431) | |
| 100 μL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ChIP Assay, Western Blot, Dot Blot, Immunocytochemistry/Immunofluorescence | |
| JE00-98 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction