missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3 Antibody (Immunomedics patent anti-Histone H3), Alexa Fluor™ 750, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28002AF750
This item is not returnable.
View return policy
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Histone H3 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Histone H3 | |
| 0.1 mL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Immunomedics patent anti-Histone H3 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction