missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, ac Lys9 Antibody (AH3-120) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£189.00
Specifications
| Antigen | Histone H3 (ac Lys9) |
|---|---|
| Clone | AH3-120 |
| Concentration | 2.1 mg/mL |
| Dilution | Western Blot 1 - 2 μg/mL, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:2000 |
| Applications | Western Blot |
Description
Histone H3 Monoclonal specifically detects Histone H3 in Human, Mouse, Rat, Bovine, C. elegans, Chicken, Drosophila, Xenopus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Histone H3 (ac Lys9) | |
| 2.1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| 126961 | |
| This Histone H3 [ac Lys9] antibody (AH3-120) was raised against a synthetic, acetylated histone H3 peptide (amino acids 7-20, acetyl K9) corresponding to the N-terminus of human histone H3, conjugated to KLH. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 15 kDa |
| AH3-120 | |
| Western Blot 1 - 2 μg/mL, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:2000 | |
| Monoclonal | |
| Mouse | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9ac, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| H3C14 | |
| IgG1 | |
| Affinity Purified | |
| Staining of (acetyl K9) histone H3 in immunoblotting is specifically inhibited with the immunizing peptide. There is no inhibition with non-acetylated histone H3 peptide. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title