missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, ac Lys9, ac Lys14 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB21-1081
This item is not returnable.
View return policy
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, C. elegans samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Dot Blot.
Specifications
| Histone H3 (ac Lys9, ac Lys14) | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9ac, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Rabbit | |
| 15 kDa | |
| 0.05 mg | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Human, Mouse, C. elegans | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Dot Blot | |
| 0.63 mg/mL | |
| Western Blot 1:500, Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:200, Dot Blot 0.1 ug/ml, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells | |
| Q71DI3, P84228 | |
| H3C14 | |
| This Histone H3 [ac Lys9, ac Lys14] antibody was raised against synthetic acetylated peptide surrounding Lysines 9 and 14 of human Histone H3.2 [Swiss Prot Q71DI3]. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction