missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, ac Lys4 Antibody (RM149) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26000
This item is not returnable.
View return policy
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ChIP assay (ChIP), Multiplex Immunoassay
Specifications
| Histone H3 | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Vertebrates | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, ChIP Assay, Multiplex | |
| RM149 | |
| Western Blot : 0.25 ug/ml-1 ug/ml, ELISA : 0.2 ug/mL - 1 ug/mL, Immunocytochemistry/ Immunofluorescence : 0.5 ug/ml - 2 ug/ml, Chromatin Immunoprecipitation (ChIP) : 1ug-5 ug, Multiplex Immunoassay : 0.05 ug/ml - 0.5 ug/ml | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| This Histone H3 [ac Lys4] antibody (RM149) was raised against an acetyl-peptide corresponding to the Histone H3 [ac Lys4] | |
| 100 μg | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction