missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Histone H3.3B Antibody (2D7-H1), Novus Biologicals™

Product Code. 18348507 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348507 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348507 Supplier Novus Biologicals™ Supplier No. H00003021M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Histone H3.3B Monoclonal antibody specifically detects Histone H3.3B in Bovine,Drosophila,Human,Mouse,Xenopus samples. It is validated for Gene Knock-Out,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3.3B
Applications Gene Knock-Out, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 2D7-H1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH17558
Gene Alias H3 histone, family 3A, H3 histone, family 3B (H3.3B), H3.3BH3.3A, H3F3, H3F3A, histone H3.3
Host Species Mouse
Immunogen H3F3B (AAH17558, 1 a.a. ∽ 136 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MARTKQTARKSTGGKAPRKQLATKAARKSAPSTGGVKKPHRYRPGTVALREIRRYQKSTELLIRKLPFQRLVREIAQDFKTDLRFQSAAIGALQEASEAYLVGLFEDTNLCAIHAKRVTIMPKDIQLARRIRGERA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3021
Target Species Bovine, Drosophila, Human, Mouse, Xenopus species
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.