Learn More
Histone H3.3 Rabbit, Clone: RM190, Abnova™
Rabbit monoclonal antibody raised against synthetic peptide of human Histone H3.3.
Brand: Abnova MAB12805.100ug
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene contains introns and its mRNA is polyadenylated, unlike most histone genes. The protein encoded is a replication-independent member of the histone H3 family. [provided by RefSeq]
Specifications
| Histone H3.3 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (0.2 ug/mL-1 ug/mL) Western Blot (0.5 ug/mL-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| H3.3A/H3F3/MGC87782/MGC87783 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3020 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| RM190 | |
| Rabbit monoclonal antibody raised against synthetic peptide of human Histone H3.3. | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| H3F3A | |
| A synthetic peptide corresponding to Histone H3.3. | |
| 100 μg | |
| Primary | |
| This antibody reacts to Histone H3.3, independent of post-translational modifications. No cross reactivity with Histone H3.1 or other histone proteins. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.