Learn More
Histone H3.3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13754-1-AP-150UL
 View more versions of this product
Product Code. 16832134
  • £349.00 / 150µL
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is located separately from the other H3 genes that are in the histone gene cluster on chromosome 6p22-p21.3.
Specifications
Specifications
Histone H3.3
Polyclonal
Unconjugated
H3F3A
H3 histone, family 3A, H3.3A, H3.3B, H3F3, H3F3A, H3F3B, Histone H3.3
Rabbit
Antigen Affinity Chromatography
RUO
15078, 289314, 3020
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P84243, P84244
H3f3a
Histone H3.3 Fusion Protein Ag4696
150 μL
Primary
Human, Mouse, Rat