missing translation for 'onlineSavingsMsg'
Learn More

Histone H3.3 Rabbit anti-Human, Mouse, Rat, Clone: S04-5J3, Novus Biologicals™

Product Code. 18385202
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18385202 25 μg 25µL
18394914 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18385202 Supplier Bio-Techne Supplier No. NBP31489425UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Histone H3.3 Monoclonal antibody specifically detects Histone H3.3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3.3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S04-5J3
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias H3.3A, H3F3, H3F3A, Histone H3.3, Histone H3.3A, Histone H3.3B, MGC87782, MGC87783
Host Species Rabbit
Immunogen A synthetic peptide of human Histone H3.3 (Uniprot#: P84243)
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 3020
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.