missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3.3 Antibody (RM190) [Janelia Fluor™ 669], Novus Biologicals™
Description
Histone H3.3 Monoclonal antibody specifically detects Histone H3.3 in Vertebrate samples. It is validated for ELISA,Western Blot,Multiplex Immunoassay
Specifications
Specifications
| Antigen | Histone H3.3 |
| Applications | ELISA, Western Blot, Multiplex |
| Classification | Monoclonal |
| Clone | RM190 |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | H3.3A, H3F3, H3F3A, Histone H3.3, Histone H3.3A, Histone H3.3B, MGC87782, MGC87783 |
| Host Species | Rabbit |
| Immunogen | A peptide corresponding to Human Histone H3.3 |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?