missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3.1 [p Ser10] Antibody (rHH3/9924), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44626-100ug
This item is not returnable.
View return policy
Description
Histone H3.1 Monoclonal antibody specifically detects Histone H3.1 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Histone H3.1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| rHH3/9924 | |
| Unconjugated | |
| H3 histone family, member D, H3.1HIST1H3D, H3/d, H3FA, H3FB, H3FC, H3FDHIST1H3F, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3A, HIST1H3B, HIST1H3C, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3e, histone cluster 1, H3e, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l | |
| A synthetic peptide corresponding to (ARK-pS-TGGKAPRKQLc) of Phosphohistone H3 (phospho S10) (Uniprot: P68431) | |
| 100 μg | |
| Epigenetics, Phospho Specific | |
| 8353 | |
| Store at 4⪚C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction