missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B, ubiquitylated Lys123 Antibody (C.15200232), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59149
This item is not returnable.
View return policy
Description
Histone H2B Monoclonal specifically detects Histone H2B in Human, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Specifications
| Histone H2B (ubiquitylated Lys123) | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| HIST2H2BF | |
| The exact immunogen is propietary information. | |
| 50 μg | |
| Chromatin Research, Epigenetics, Histones and Modified Histones | |
| 440689 | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| Western Blot, ChIP Assay | |
| C.15200232 | |
| Western Blot 1:500, Chromatin Immunoprecipitation (ChIP) 2 ug/IP | |
| FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Yeast | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction