missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B Antibody (Immunomedics patent anti-Histone H2B), mFluor Violet 610 SE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28001MFV610
This item is not returnable.
View return policy
Description
Histone H2B Monoclonal antibody specifically detects Histone H2B in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Histone H2B | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
| Histone H2B | |
| 0.1 mL | |
| Chromatin Research, Epigenetics | |
| 440689 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Immunomedics patent anti-Histone H2B | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction