missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Histone H2B Antibody (Immunomedics patent anti-Histone H2B) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | Histone H2B |
|---|---|
| Clone | Immunomedics patent anti-Histone H2B |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
30048573
|
Novus Biologicals™
NBP3-28001-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045462
|
Novus Biologicals™
NBP3-28001-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30048251
|
Novus Biologicals™
NBP3-28001-100ug |
100 μg |
£257.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H2B Monoclonal antibody specifically detects Histone H2B in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpécification
| Histone H2B | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Chromatin Research, Epigenetics | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
| Histone H2B | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Immunomedics patent anti-Histone H2B | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P33778 | |
| 440689 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit