missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B (acetyl K23) Rabbit anti-Human, Clone: RM260, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB15113.100ug
This item is not returnable.
View return policy
Description
Rabbit monoclonal antibody raised against synthetic acetyl peptide of human histone H2B (acetyl K23).
- High specificity
- High reactivity
Specifications
| Histone H2B (acetyl K23) | |
| Monoclonal | |
| Unconjugated | |
| ELISA (0.2-1 ug/mL) Immunocytochemistry (0.5-2 ug/mL) Multiplex (0.1-0.5 ug/mL) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| P33778 | |
| HIST1H2BB | |
| A synthetic acetyl peptide corresponding to residues surrounding K23 of human histone H2B. | |
| 100 μg | |
| Primary | |
| This antibody reacts to histone H2B acetylated at K23. No cross reactivity with other acetylated lysines in histones. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunocytochemistry, Western Blot | |
| RM260 | |
| Rabbit monoclonal antibody raised against synthetic acetyl peptide of human histone H2B (acetyl K23). | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| H2B.1/H2B/f/H2BFF/MGC119804 | |
| Rabbit | |
| Protein A affinity purification | |
| RUO | |
| 3018 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction