missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Histone H2B Monoclonal antibody specifically detects Histone H2B in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | Histone H2B |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Clone | GT578 |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 |
| Host Species | Mouse |
| Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Histone H2B (acetyl Lys20). The exact sequence is proprietary. |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?