missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AY/macroH2A.1 Antibody (RM248), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-61494
This item is not returnable.
View return policy
Description
Histone H2AY/macroH2A.1 Monoclonal antibody specifically detects Histone H2AY/macroH2A.1 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay
Specifications
| Histone H2AY/macroH2A.1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5 μg/mL - 2 μg/mL, ELISA 0.2 μg/mL - 1 μg/mL, Immunocytochemistry/ Immunofluorescence 1 μg/mL - 2 μg/mL, Multiplex Immunoassay 0.2 μg/mL - 1 μg/mL | |
| core histone macro-H2A.1, H2A histone family, member Y, H2A.y, H2A/y, H2AF12M, H2AFJ, Histone H2A.y, Histone macroH2A1, histone macroH2A1.1, histone macroH2A1.2, MACROH2A1, MACROH2A1.1, macroH2A1.2, Medulloblastoma antigen MU-MB-50.205, mH2A1 | |
| A peptide corresponding to the C-terminus of Human Histone H2AY/macroH2A.1 | |
| 100 μg | |
| Primary | |
| Human, Vertebrates | |
| Purified |
| Western Blot, ELISA, Immunofluorescence, Multiplex | |
| RM248 | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 9555 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction