missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AY/macroH2A.1 Antibody (RM248), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26031CL1
This item is not returnable.
View return policy
Description
Histone H2AY/macroH2A.1 Monoclonal antibody specifically detects Histone H2AY/macroH2A.1 in Human,Vertebrate samples. It is validated for ELISA,Western Blot,Multiplex Immunoassay,Immunocytochemistry/Immunofluorescence
Specifications
| Histone H2AY/macroH2A.1 | |
| Monoclonal | |
| CoraFluor 1 | |
| core histone macro-H2A.1, H2A histone family, member Y, H2A.y, H2A/y, H2AF12M, H2AFJ, Histone H2A.y, Histone macroH2A1, histone macroH2A1.1, histone macroH2A1.2, MACROH2A1, MACROH2A1.1, macroH2A1.2, Medulloblastoma antigen MU-MB-50.205, mH2A1 | |
| A peptide corresponding to the C-terminus of Human Histone H2AY/macroH2A.1 | |
| 0.1 mL | |
| Primary | |
| Human, Vertebrates | |
| Purified |
| ELISA, Western Blot, Multiplex, Immunocytochemistry/Immunofluorescence | |
| RM248 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 9555 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction