missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AX Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Histone H2AX Polyclonal specifically detects Histone H2AX in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Histone H2AX |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Host Species | Rabbit |
| Immunogen | Recombinant protein of human Histone H2AX. |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?