missing translation for 'onlineSavingsMsg'
Learn More

Histone H2AX, p Ser139 Antibody (RM224), Novus Biologicals™

Product Code. 18339614 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339614 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339614 Supplier Novus Biologicals Supplier No. NBP318533

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Histone H2AX Monoclonal antibody specifically detects Histone H2AX in Human, Vertebrates samples. It is validated for Western Blot, ELISA, Immunofluorescence, Multiplex Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2AX
Applications Western Blot, ELISA, Immunofluorescence, Peptide Array
Classification Monoclonal
Clone RM224
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5 to 2 μg/mL, ELISA 0.2 to 1 μg/mL, Immunocytochemistry/Immunofluorescence 0.5 to 2 μg/mL, Multiplex Immunoassay 0.1 to 1 μg/mL
Formulation 50% Glycerol, PBS, 1% BSA
Gene Alias H2A.X, H2A/X, H2AFX
Host Species Rabbit
Immunogen A phospho-peptide corresponding to Phospho-Histone H2AX (Ser139)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3014
Target Species Human, Vertebrates
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.