missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AX, p Ser139 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-384PE
This item is not returnable.
View return policy
Description
Histone H2AX (p Ser139) Polyclonal antibody specifically detects Histone H2AX (p Ser139) in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry, Immunoassay.
Specifications
| Histone H2AX (p Ser139) | |
| Polyclonal | |
| Flow Cytometry, Knockout Validated | |
| H2A.X, H2A/X, H2AFX | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG |
| Flow Cytometry, Immunoassay | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| H2AFX | |
| This Histone H2AX [p Ser139] Antibody was developed against to a region surrounding phosphorylated serine 139 of human histone H2AX [Swiss-Prot entry P16104] (GeneID 3014). | |
| 0.1 mL | |
| Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific | |
| 3014 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction