missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AX Antibody (CL5157) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21157JF669
This item is not returnable.
View return policy
Description
Histone H2AX Monoclonal antibody specifically detects Histone H2AX in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Histone H2AX | |
| Monoclonal | |
| Janelia Fluor 669 | |
| H2A.X, H2A/X, H2AFX | |
| This antibody was developed using a synthetic peptide derived from P16104, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific | |
| 3014 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL5157 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction