missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 4 beta 7/LPAM-1 Antibody (CL11293) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21189F
This item is not returnable.
View return policy
Description
Integrin alpha 4 beta 7/LPAM-1 Monoclonal antibody specifically detects Integrin alpha 4 beta 7/LPAM-1 in Human samples. It is validated for Western Blot
Specifications
| Integrin alpha 4 beta 7/LPAM-1 | |
| Monoclonal | |
| FITC | |
| 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
| This antibody was developed using a synthetic peptide derived from P13612, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Innate Immunity | |
| 3676 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL11293 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction