missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2A.Z Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54618
This item is not returnable.
View return policy
Description
Histone H2A.Z Polyclonal antibody specifically detects Histone H2A.Z in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ChIP assay (ChIP), ChIP assay Sequencing
Specifications
| Histone H2A.Z | |
| Polyclonal | |
| Western Blot 1:1000, ELISA 1:5000, Immunocytochemistry/ Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 0.5-1 μg/IP, Chromatin Immunoprecipitation Sequencing | |
| H2A histone family, member Z, H2A.z, H2A/z, H2AZ histone, H2AZMGC117173, histone H2A.Z | |
| H2A.Z | |
| 50 μg | |
| Epigenetics | |
| 3015 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence, ChIP Assay, ChIP sequencing (ChIP-seq) | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction