missing translation for 'onlineSavingsMsg'
Learn More

Histone H2A.Z, ac Lys7, ac Lys11, ac Lys4 Antibody, Novus Biologicals™

Product Code. 18398893 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398893 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398893 Supplier Novus Biologicals Supplier No. NBP318689

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Histone H2A.Z Polyclonal antibody specifically detects Histone H2A.Z in Human samples. It is validated for Western Blot, ChIP assay, ELISA, Immunofluorescence, Dot Blot, ChIP assay Sequencing
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2A.Z
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Chromatin Immunoprecipitation 0.5 ug/ChIP, ELISA 1:5000, Immunocytochemistry/Immunofluorescence 1:500, Dot Blot 1:20000, Chromatin Immunoprecipitation Sequencing 0.5 ug/ChIP
Formulation PBS
Gene Accession No. P0C0S5
Gene Alias H2A histone family, member Z, H2A.z, H2A/z, H2AZ histone, H2AZMGC117173, histone H2A.Z
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against Histone H2A.Z acetylated at lysines 4, 7 and 11, using a KLHconjugated synthetic peptide.
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 3015
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.