missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Histone H1T Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566983
This item is not returnable.
View return policy
Description
Immunogen sequence: SETVPAASASA GVAAMEKLPT KKRGRKPAGL ISASRKVPNL SVSKLITEAL SVSQERVGMS Highest antigen sequence identity to the following orthologs - mouse 55%, rat 57%.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element.
Specifications
| Histone H1T | |
| Polyclonal | |
| Unconjugated | |
| H1-6 | |
| dJ221C16.2; H1 histone family, member T (testis-specific); H1.6; H1ft; H1T; HIST1H1T; histone 1, H1t; histone cluster 1, H1t; histone H1t; Testicular H1 histone | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P22492 | |
| H1-6 | |
| Recombinant Human Histone H1T. Recombinant protein control fragment (Product #RP-107626). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu