missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Histone H1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530055
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, HepG2 nuclear extract, mouse liver. Predicted reactivity: Mouse (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This locus is the ortholog of a mouse protein-coding gene that displays characteristics of a linker histone and is expressed in nuclei of late maturing spermatids. The human locus is expressed; however, the open reading frame has been disrupted by a frameshift so it is unlikely to encode a functional protein.
Specifications
| Histone H1 | |
| Polyclonal | |
| Unconjugated | |
| H1-2 | |
| 0610008C09Rik; CTL-1; H1; H1 antibody; H1 histone family member 0; H1 histone family, member 0; H1 histone family, member 1; H1 histone family, member 2; H1 histone family, member 5; H1 VAR.1; H1 VAR.3; H1 VAR.5; H1(0); H1.0; H1.0, H1(0), H1-0; H1.1; H1.12 linker histone, cluster member; H1.2; H1.2 linker histone, cluster member; H1.5; H1.9; H10; H1-0; H1-1; H1-12; H1-2; H1A; H1B; H1c; H1F0; H1f1; H1F2; H1F3; H1F5; H1FV; H1s-1; H1s-3; H1var1; H1var3; Hils1; His1a; HIST1; HIST1H1A; HIST1H1B; HIST1H1B antibody; HIST1H1C; HIST1H1D; HIST1H1E; Histone 1 antibody; histone 1, family 5; histone 1, H1a; histone 1, H1b; histone 1, H1c; histone cluster 1, H1a; histone cluster 1, H1b; histone cluster 1, H1c; histone cluster 1, H1e; histone H1; histone H1'; Histone H1(0); histone H1.0; Histone H1.0, N-terminally processed; Histone H1.1; histone H1.2; Histone H1.3; Histone H1.5; histone H1a; Histone H1b; histone H1c; Histone H1d; histone H1-like protein in spermatids 1; histone H1s-1; Histone H1s-2; Histone H1s-3; histone linker H1 domain, spermatid-specific 1 (pseudogene); spermatid-specific linker histone H1-like protein; TISP64 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116883558, 14958, 24437, 291145, 3005, 3006, 3009, 3024, 50708, 56702, 680522, 80838 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.53 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| D3ZBN0, D4A3K5, P07305, P10922, P15864, P16401, P16403, P43275, P43276, P43278, Q02539 | |
| H1-0, H1-1, H1-2, H1-5 | |
| Recombinant protein encompassing a sequence within the center region of human Histone H1.0. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction