missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1 Antibody (derlotuximab), DyLight 650, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27977C
This item is not returnable.
View return policy
Description
Histone H1 Monoclonal antibody specifically detects Histone H1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Histone H1 | |
| Monoclonal | |
| DyLight 650 | |
| H1 histone family, member 1, H1.1, H1A, H1F1HIST1, histone 1, H1a, histone cluster 1, H1a, histone H1.1, MGC126642, MGC138345 | |
| Histone H1 | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Epigenetics, Nuclear Envelope Markers | |
| 3024 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| derlotuximab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction