missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1.4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-16044-100UL
This item is not returnable.
View return policy
Description
Histone H1.4 Polyclonal antibody specifically detects Histone H1.4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Histone H1.4 | |
| Polyclonal | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| dJ221C16.5, H1 histone family, member 4, H1.4, H1F4H1E, histone 1, H1e, histone cluster 1, H1e, histone H1.4, Histone H1b, MGC116819 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Histone H1.4 (NP_005312.1). | |
| 100 μg | |
| Epigenetics | |
| 3008 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction