missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1.4, p Thr17 Antibody (3E1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26420-100ul
This item is not returnable.
View return policy
Description
Histone H1.4 Monoclonal antibody specifically detects Histone H1.4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry
Specifications
| Histone H1.4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry | |
| 3E1 | |
| ELISA, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:30-1:200 | |
| dJ221C16.5, H1 histone family, member 4, H1.4, H1F4H1E, histone 1, H1e, histone cluster 1, H1e, histone H1.4, Histone H1b, MGC116819 | |
| Synthesized peptide [UniProt P10412] | |
| 100 μL | |
| Epigenetics | |
| 3008 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction