missing translation for 'onlineSavingsMsg'
Learn More

Histone H1.3 Rabbit anti-Human, Mouse, Rat, Clone: 3K5C10, Novus Biologicals™

Product Code. 18339716 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339716 100 μg 100µL
18357776 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18339716 Supplier Novus Biologicals Supplier No. NBP316927100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Histone H1.3 Monoclonal antibody specifically detects Histone H1.3 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H1.3
Applications Western Blot
Classification Monoclonal
Clone 3K5C10
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias H1 histone family, member 3, H1.3, H1F3H1D, histone 1, H1d, histone cluster 1, H1d, histone H1.3, Histone H1c, MGC138176
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around T17 of human Histone H1.3 (P16402).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 3007
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.