missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Histone H1.3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535892
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6. [provided by RefSeq].
Specifications
| Histone H1.3 | |
| Polyclonal | |
| Unconjugated | |
| H1-3 | |
| H1 histone family, member 3; H1 VAR.4; H1.3; H1D; H1f3; H1s-2; H1s-4; Hist1h1d; histone 1, family 3; histone 1, H1d; histone cluster 1, H1d; histone H1.3; Histone H1c; histone H1s-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14957, 3007 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P16402, P43277 | |
| H1-3, HIST1H1D | |
| Recombinant protein encompassing a sequence within the center region of human Histone H1.3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction