missing translation for 'onlineSavingsMsg'
Learn More

Histone H1.1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£292.00 - £488.00

Specifications

Antigen Histone H1.1
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18470732
View Documents
Novus Biologicals
NBP2-33402-25ul
25 μL
£292.00
25µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
18179103
missing translation for 'viewDocuments'
Novus Biologicals
NBP2-33402
0.1 mL
£488.00
0.10mL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Histone H1.1 Polyclonal specifically detects Histone H1.1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Histone H1.1
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
Q02539
3024
This antibody was developed against a recombinant protein corresponding to amino acids: SVETKPGASKVATKTKATGASKKLKKATGASKKSVKTPK
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Polyclonal
Rabbit
Cellular Markers, Chromatin Research, Epigenetics, Histones and Modified Histones, Nuclear Envelope Markers
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
H1 histone family, member 1, H1.1, H1A, H1F1HIST1, histone 1, H1a, histone cluster 1, H1a, histone H1.1, MGC126642, MGC138345
H1-1
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.